Merck
  • [Mediated role of NO-syntase, protein kinase C and KATP-channels in realization of cardioprotective impact of cannabinoid HU-210].
[Mediated role of NO-syntase, protein kinase C and KATP-channels in realization of cardioprotective impact of cannabinoid HU-210].
Eksperimental'naia i klinicheskaia farmakologiia (2012-01-01)
L N Maslov, O V Lasukova, A V Krylatov, L Khanush, Iu B Lishmanov
ABSTRACT
It was shown that perfusion of the isolated heart of rat with solution containing the CB1- and CB2-receptor agonist HU-210 at concentrations of 0.1 or 1.0 microM/L for a duration of 10 min at 20 min before global ischemia (45 min) and reperfusion (30 min) promotes a twofold decrease in creatine kinase levels in coronary effluent. It was established that KATP channel blockade by glibenclamide (1 microM/L) or inhibition of protein kinase C (2 microM/L) by chelerythrine abolishes the cardioprotective effect of HU-210. The inhibitor of NO synthase L-NAME (1 microM/L) had no effect on the anti-necrotic effect of HU-210. Thus, the intracellular signaling mechanism of the cardioprotective effect of the CB-agonist HU-210 involves the activation of KATP channels and protein kinase C without the participation of NO-synthase.
Product Description
Supelco
HU-210 solution, 100 μg/mL in methanol, ampule of 1 mL, (Spice Cannabinoid), certified reference material, Cerilliant®
Sigma-Aldrich
HU-210, solid (air sensitive)