Merck
  • A novel and simple procedure to synthesize chitosan-graft-polycaprolactone in an ionic liquid.
A novel and simple procedure to synthesize chitosan-graft-polycaprolactone in an ionic liquid.
Carbohydrate polymers (2013-04-03)
Zhaodong Wang, Liuchun Zheng, Chuncheng Li, Dong Zhang, Yaonan Xiao, Guohu Guan, Wenxiang Zhu
ABSTRACT
An ionic liquid, 1-ethyl-3-methylimidazolium acetate (EMIMAc), was synthesized and employed as a homogeneous and green reaction media to prepare chitosan-graft-polycaprolactone (CS-g-PCL) via ring-opening polymerization, using stannous octoate (Sn(Oct)2) as a catalyst. The structures and compositions of copolymers could be facilely controlled by the reaction conditions and feed ratios. The grafting content of polycaprolactone (PCL) could reach as high as 630%. The chemical structures of the copolymers were systematically characterized by (1)H NMR, Fourier transform infrared spectroscopy (FTIR) and wide-angle X-ray diffraction (WAXD), while thermal properties were studied by thermogravimetric analysis (TGA) and differential scanning calorimetry (DSC). The thermal stability and glass transition temperature (Tg) of the graft copolymers vary regularly with the change of PCL grafting content.
Sigma-Aldrich
Polycaprolactone, average Mn 45,000
Sigma-Aldrich
Polycaprolactone, average Mn 80,000
Sigma-Aldrich
Polycaprolactone, average Mw ~14,000, average Mn ~10,000 by GPC