Merck
  • Simultaneous determination of trimipramine and its major metabolites by high-performance liquid chromatography.
Simultaneous determination of trimipramine and its major metabolites by high-performance liquid chromatography.
Journal of chromatography (1991-05-03)
A A Gulaid, G A Jahn, C Maslen, M J Dennis
ABSTRACT
Trimipramine is a tricyclic antidepressant drug often assayed by gas chromatographic or gas chromatographic-mass spectrometry techniques. A high-performance liquid chromatographic method with electrochemical detection is described for the assay of trimipramine and its major metabolites, monodesmethyltrimipramine and 2-hydroxytrimipramine, in plasma. The method is sensitive, accurate and robust and thus suitable for routinely assaying samples following single doses of trimipramine to man. The assay was applied to plasma samples obtained following a single 50-mg dose of trimipramine to healthy volunteers.
Product Description
Supelco
N-Desmethyltrimipramine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®