Merck
  • Substances derived from 4-ethyl octanoic acid account for primer pheromone activity for the "male effect" in goats.
Substances derived from 4-ethyl octanoic acid account for primer pheromone activity for the "male effect" in goats.
The Journal of veterinary medical science (2003-10-09)
Eri Iwata, Takefumi Kikusui, Yukari Takeuchi, Yuji Mori
ABSTRACT
A major constituent of the characteristic "goaty odor" 4-ethyl octanoic acid (4EOA) was previously shown to have no primer pheromone activity. This was also confirmed by our own bioassay system utilizing the recording technique of neural activity of the hypothalamic gonadotropin-releasing hormone pulse generator in goats. However, when the synthetic 4EOA solution was kept at room temperature for several months, primer pheromone activity appeared in the same solution. Headspace gas chromatography/mass spectrometry analysis revealed that there were several newly formed substances in addition to 4EOA samples with primer pheromone activity. These results suggest that 4EOA derived substance(s) but not 4EOA itself is(are) primer pheromone in goats.
Sigma-Aldrich
4-Ethyloctanoic acid, ≥98%, FG
Pricing and availability is not currently available.