Merck
  • Simultaneous quantification of insecticides including carbaryl in drinking water by gas chromatography using dual electron-capture and nitrogen-phosphorus detection.
Simultaneous quantification of insecticides including carbaryl in drinking water by gas chromatography using dual electron-capture and nitrogen-phosphorus detection.
Journal of chromatography. A (1997-05-09)
Y S Oh-Shin, M Ko, H S Shin
ABSTRACT
A rapid and simple gas chromatographic method for the simultaneous determination of malathion, parathion, fenitrothion, diazinon and carbaryl in drinking water is presented. A fused-silica SE-54 column was used for the separation of the insecticides and was split into two specific detectors; electron-capture and nitrogen-phosphorus detectors by using column switching. A water sample was extracted with methylene chloride. The organic phase was evaporated and the residue was derivatized with pentafluoropropionic acid anhydride. The detection limit was below 0.1 ng/ml and the calibration curves showed good linearity with r = 0.998 approximately 1.000. The method was sensitive, reproducible and simple enough to permit the reliable routine analysis of the pesticides in drinking water.
Product Description
Supelco
Pentafluoropropionic anhydride, for GC derivatization, LiChropur, 99%
Sigma-Aldrich
Pentafluoropropionic anhydride, 99%
Sigma-Aldrich
Pentafluoropropionic anhydride, purum, ≥97.0% (GC)