Merck
  • Interleukin-4 and prostaglandin E2 synergistically up-regulate 3β-hydroxysteroid dehydrogenase type 2 in endometrioma stromal cells.
Interleukin-4 and prostaglandin E2 synergistically up-regulate 3β-hydroxysteroid dehydrogenase type 2 in endometrioma stromal cells.
The Journal of clinical endocrinology and metabolism (2013-03-02)
Yoko Urata, Yutaka Osuga, Ikumi Akiyama, Miwako Nagai, Gentaro Izumi, Masashi Takamura, Akiko Hasegawa, Miyuki Harada, Tetsuya Hirata, Yasushi Hirota, Osamu Yoshino, Kaori Koga, Shiro Kozuma
ABSTRACT
Endometriosis is a chronic inflammatory disease in which immune response and production of estrogen in endometriotic tissues are involved in the development of the disease. Prostaglandin E2 (PGE2) stimulates aromatase (P450arom) expression in endometrioma stromal cells (ESCs) and increases the production of estrogens. On the other hand, an accumulating amount of evidence suggests that IL-4, a typical Th2 cytokine, plays important roles in the disease. The objective of the investigation was to study the effect of IL-4 on the expression of 3β-hydroxysteroid dehydrogenase (HSD3B2), a pivotal enzyme for estrogen production, in ESCs. ESCs were isolated from ovarian endometrioma tissues and cultured with IL-4 and PGE2. CP-690550, a Janus protein tyrosine kinase 3 inhibitor, and HSD3B2 small interfering RNA were added to the culture. Gene expression of HSD3B2 and P450arom was examined by quantitative RT-PCR. Dehydroepiandrosterone (DHEA) was added to the culture, and then the combined enzyme activity of HSD3B2, which converts DHEA to androstenedione, and P450arom, which converts androstenedione to estrone, was examined by measuring estrone concentration in the supernatants with a specific enzyme immunoassay. IL-4 increased the expression of HSD3B2 mRNA in a dose-dependent manner. CP-650550 inhibited the IL-4-induced increase in HSD3B2 mRNA expression. PGE2 also increased the expression of HSD3B2 mRNA, and the combination of IL-4 and PGE2 synergistically increased the expression of HSD3B2 mRNA. IL-4 had no effect on the expression of P450arom mRNA, whereas PGE2 increased the expression of P450arom mRNA. Although PGE2 alone increased the production of estrone from DHEA, the combination of IL-4 and PGE2 significantly augmented the production of estrone from DHEA. The enhanced production of estrone by the combination of IL-4 and PGE2 was inhibited by CP-690550 and HSD3B2 small interfering RNA. IL-4 in combination with PGE2 may enhance estrogen production in endometriotic tissues, implying an elaborate mechanism that Th2 immune response augments inflammation-dependent progression of the disease.
Sigma-Aldrich
Estrone, ≥99%
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Estrone, European Pharmacopoeia (EP) Reference Standard