Merck
  • Iridium-catalyzed selective α-alkylation of unactivated amides with primary alcohols.
Iridium-catalyzed selective α-alkylation of unactivated amides with primary alcohols.
Organic letters (2013-02-16)
Le Guo, Yinghua Liu, Wubing Yao, Xuebing Leng, Zheng Huang
ABSTRACT
The first α-alkylation of unactivated amides with primary alcohols is described. An effective and robust iridium pincer complex has been developed for selective α-alkylation of tertiary and secondary acetamides involving a "borrowing hydrogen" methodology. The method is compatible with alcohols bearing various functional groups. This presents a convenient and environmentally benign protocol for α-alkylation of amides.
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis