Merck
  • Hypochlorous acid turn-on fluorescent probe based on oxidation of diphenyl selenide.
Hypochlorous acid turn-on fluorescent probe based on oxidation of diphenyl selenide.
Organic letters (2013-02-05)
Shi-Rong Liu, Shu-Pao Wu
ABSTRACT
A BODIPY-based fluorescent probe, HCSe, has been successfully developed for the rapid detection of hypochlorous acid based on the specific HOCl-promoted oxidation of diphenyl selenide in response to the amount of HOCl. Confocal fluorescence microscopy imaging using RAW264.7 cells showed that the new probe HCSe could be used as an effective fluorescent probe for detecting HOCl in living cells.
Sigma-Aldrich
Biphenyl, ≥99%
Supelco
Biphenyl, PESTANAL®, analytical standard
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%