Merck
  • Hole-transporting transistors and circuits based on the transparent inorganic semiconductor copper(I) thiocyanate (CuSCN) processed from solution at room temperature.
Hole-transporting transistors and circuits based on the transparent inorganic semiconductor copper(I) thiocyanate (CuSCN) processed from solution at room temperature.
Advanced materials (Deerfield Beach, Fla.) (2013-01-03)
Pichaya Pattanasattayavong, Nir Yaacobi-Gross, Kui Zhao, Guy Olivier Ngongang Ndjawa, Jinhua Li, Feng Yan, Brian C O'Regan, Aram Amassian, Thomas D Anthopoulos
ABSTRACT
The wide bandgap and highly transparent inorganic compound copper(I) thiocyanate (CuSCN) is used for the first time to fabricate p-type thin-film transistors processed from solution at room temperature. By combining CuSCN with the high-k relaxor ferroelectric polymeric dielectric P(VDF-TrFE-CFE), we demonstrate low-voltage transistors with hole mobilities on the order of 0.1 cm(2) V(-1) s(-1) . By integrating two CuSCN transistors, unipolar logic NOT gates are also demonstrated.
Sigma-Aldrich
Copper(I) thiocyanate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cuprous thiocyanate, 35 mg/mL in diethyl sulfide
Pricing and availability is not currently available.