Merck
  • Systematic review with dose-response meta-analyses between vitamin B-12 intake and European Micronutrient Recommendations Aligned's prioritized biomarkers of vitamin B-12 including randomized controlled trials and observational studies in adults and elderly persons.
Systematic review with dose-response meta-analyses between vitamin B-12 intake and European Micronutrient Recommendations Aligned's prioritized biomarkers of vitamin B-12 including randomized controlled trials and observational studies in adults and elderly persons.
The American journal of clinical nutrition (2012-12-28)
Carla Dullemeijer, Olga W Souverein, Esmée L Doets, Hilko van der Voet, Janneke P van Wijngaarden, Waldo J de Boer, Maria Plada, Rosalie A M Dhonukshe-Rutten, Paulette H In 't Veld, Adrienne E J M Cavelaars, Lisette C P G M de Groot, Pieter van 't Veer
ABSTRACT
Many randomized controlled trials (RCTs) and observational studies have provided information on the association between vitamin B-12 intake and biomarkers. The use of these data to estimate dose-response relations provides a useful means to summarize the body of evidence. We systematically reviewed studies that investigated vitamin B-12 intake and biomarkers of vitamin B-12 status and estimated dose-response relations with the use of a meta-analysis. This systematic review included all RCTs, prospective cohort studies, nested case-control studies, and cross-sectional studies in healthy adult populations published through January 2010 that supplied or measured dietary vitamin B-12 intake and measured vitamin B-12 status as serum or plasma vitamin B-12, methylmalonic acid (MMA), or holotranscobalamin. We calculated an intake-status regression coefficient ( ) for each individual study and calculated the overall pooled and SE ( ) by using random-effects meta-analysis on a double-log scale. The meta-analysis of observational studies showed a weaker slope of dose-response relations than the meta-analysis of RCTs. The pooled dose-response relation of all studies between vitamin B-12 intake and status indicated that a doubling of the vitamin B-12 intake increased vitamin B-12 concentrations by 11% (95% CI: 9.4%, 12.5%). This increase was larger for studies in elderly persons (13%) than in studies in adults (8%). The dose-response relation between vitamin B-12 intake and MMA concentrations indicated a decrease in MMA of 7% (95% CI: -10%, -4%) for every doubling of the vitamin B-12 intake. The assessment of risk of bias within individual studies and across studies indicated risk that was unlikely to seriously alter these results. The obtained dose-response estimate between vitamin B-12 intake and status provides complementary evidence to underpin recommendations for a vitamin B-12 intake of populations.
Sigma-Aldrich
Methylmalonic acid, 99%
Supelco
Methylmalonic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®