Merck
  • Determination of tamsulosin in human plasma by liquid chromatography/tandem mass spectrometry and its application to a pharmacokinetic study.
Determination of tamsulosin in human plasma by liquid chromatography/tandem mass spectrometry and its application to a pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-11-17)
Chang-Ik Choi, Hye-In Lee, Jung-Woo Bae, Yun-Jeong Lee, Ji-Yeong Byeon, Choon-Gon Jang, Seok-Yong Lee
ABSTRACT
Tamsulosin, a selective α₁-adrenoceptor antagonist, is used for the treatment of benign prostatic hyperplasia (BPH). We developed and validated a rapid, sensitive, and simplified liquid chromatography analytical method utilizing tandem mass spectrometry (LC-MS/MS) for the determination of tamsulosin in human plasma. After liquid-liquid extraction with methyl t-butyl ether, chromatographic separation of tamsulosin was achieved using a reversed-phase Luna C₁₈ column (2.0 mm × 50 mm, 5 μm particles) with a mobile phase of 10 mM ammonium formate buffer (pH 3.5)-methanol (25:75, v/v) and quantified by MS/MS detection in ESI positive ion mode. The flow rate of the mobile phase was 200 μL/min and the retention times of tamsulosin and the internal standard (IS, diphenhydramine) were 0.8 and 0.9 min, respectively. The calibration curves were linear over a range of 0.01-20 ng/mL (r>0.999). The lower limit of quantification using 500 μL of human plasma was 0.01 ng/mL. The mean accuracy and precision for intra- and inter-day validation of tamsulosin were both within acceptable limits. The present LC-MS/MS method showed improved sensitivity for quantification of tamsulosin in human plasma compared with previously described analytical methods. The validated method was successfully applied to a pharmacokinetic study in humans.
Product Description
Supelco
Diphenhydramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Diphenhydramine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard