Merck
  • Evolutionarily repurposed networks reveal the well-known antifungal drug thiabendazole to be a novel vascular disrupting agent.
Evolutionarily repurposed networks reveal the well-known antifungal drug thiabendazole to be a novel vascular disrupting agent.
PLoS biology (2012-08-29)
Hye Ji Cha, Michelle Byrom, Paul E Mead, Andrew D Ellington, John B Wallingford, Edward M Marcotte
ABSTRACT
Studies in diverse organisms have revealed a surprising depth to the evolutionary conservation of genetic modules. For example, a systematic analysis of such conserved modules has recently shown that genes in yeast that maintain cell walls have been repurposed in vertebrates to regulate vein and artery growth. We reasoned that by analyzing this particular module, we might identify small molecules targeting the yeast pathway that also act as angiogenesis inhibitors suitable for chemotherapy. This insight led to the finding that thiabendazole, an orally available antifungal drug in clinical use for 40 years, also potently inhibits angiogenesis in animal models and in human cells. Moreover, in vivo time-lapse imaging revealed that thiabendazole reversibly disassembles newly established blood vessels, marking it as vascular disrupting agent (VDA) and thus as a potential complementary therapeutic for use in combination with current anti-angiogenic therapies. Importantly, we also show that thiabendazole slows tumor growth and decreases vascular density in preclinical fibrosarcoma xenografts. Thus, an exploration of the evolutionary repurposing of gene networks has led directly to the identification of a potential new therapeutic application for an inexpensive drug that is already approved for clinical use in humans.
Product Description
Supelco
Thiabendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Thiabendazole, PESTANAL®, analytical standard
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Sigma-Aldrich
Thiabendazole, suitable for plant cell culture
Tiabendazole, European Pharmacopoeia (EP) Reference Standard