Merck
  • Finely dispersed single-walled carbon nanotubes for polysaccharide hydrogels.
Finely dispersed single-walled carbon nanotubes for polysaccharide hydrogels.
ACS applied materials & interfaces (2012-08-23)
Liang Yu Yan, Hailan Chen, Peng Li, Dong-Hwan Kim, Mary B Chan-Park
ABSTRACT
Here we demonstrate a polysaccharide hydrogel reinforced with finely dispersed single-walled carbon nanotubes (SWNTs) using biocompatible dispersants O-carboxymethylchitosan (OC) and chondroitin sulfate A (CS-A) as a structural support. Both of the dispersants can disperse SWNTs in aqueous solutions and hydrogel matrix as individual tubes or small bundles. Additionally, we have found that compressive modulus and strain of the hydrogels reinforced with SWNTs were enhanced as much as two times by the addition of a few weight percent of SWNTs. Moreover, the SWNT-incorporated hydrogels exhibited lower impedance and higher charge capacity than the alginate/dispersant hydrogel without SWNTs. The OC and the CS-A demonstrated much higher reinforcing enhancement than a commercially available dispersant, sodium dodecyl sulfate. Combined with the experimental data on the mechanical and electrical properties, the biocompatibility of OC and CS-A can provide the possibility of biomedical application of the SWNT-reinforced hydrogels.
Sigma-Aldrich
Gluconolactone, 99.0-101.0%, meets USP testing specifications
Sigma-Aldrich
D-(+)-Gluconic acid δ-lactone, ≥99.0% (GC)