Merck
  • Solution processed MoO3 interfacial layer for organic photovoltaics prepared by a facile synthesis method.
Solution processed MoO3 interfacial layer for organic photovoltaics prepared by a facile synthesis method.
Advanced materials (Deerfield Beach, Fla.) (2012-04-11)
Seiichiro Murase, Yang Yang
ABSTRACT
An MoO(3) film spin-coated from a solution prepared by an extremely facile and cost-effective synthetic method is introduced as an anode buffer layer of bulk-heterojunction polymer photovoltaic devices. The device efficiency using the MoO(3) anode buffer layer is comparable to that using a conventional PEDOT:PSS layer without annealing at an elevated temperature.
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%