Merck
  • InCl3-driven regioselective synthesis of functionalized/annulated quinolines: scope and limitations.
InCl3-driven regioselective synthesis of functionalized/annulated quinolines: scope and limitations.
Chemistry, an Asian journal (2012-02-09)
Tanmoy Chanda, Rajiv Kumar Verma, Maya Shankar Singh
ABSTRACT
The efficient, regioselective synthesis of functionalized/annulated quinolines was achieved by the coupling of 2-aminoaryl ketones with alkynes/active methylenes/α-oxoketene dithioacetals promoted by InCl(3) in refluxing acetonitrile as well as under solvent-free conditions in excellent yields. This transformation presumably proceeded through the hydroamination-hydroarylation of alkynes, and the Friedländer annulation of active methylene compounds and α-oxoketene dithioacetals with 2-aminoarylketones. In addition, simple reductive and oxidative cyclization of 2-nitrobenzaldehyde and 2-aminobenzylalcohol, respectively, afforded substituted quinolines. Systematic optimization of the reaction parameters allowed us to identify two-component coupling (2CC) conditions that were tolerant of a wide range of functional groups, thereby providing densely functionalized/annulated quinolines. This approach tolerates the synthesis of various bioactive quinoline frameworks from the same 2-aminoarylketones under mild conditions, thus making this strategy highly useful in diversity-oriented synthesis (DOS). The scope and limitations of the alkyne-, activated methylene-, and α-oxoketene dithioacetal components on the reaction were also investigated.
Sigma-Aldrich
Indium(III) chloride, 98%
Sigma-Aldrich
Indium(III) chloride, anhydrous, powder, ≥99.999% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 99.999% trace metals basis