Merck
Catalytic hydroacylation as an approach to homoaldol products.
Organic letters (2011-11-09)
Stephen K Murphy, David A Petrone, Matthew M Coulter, Vy M Dong
ABSTRACT
A method has been developed for the intermolecular hydroacylation of homoallyl alcohols with salicylaldehydes to furnish homoaldol products in 50-98% yields. The method also applies to the hydroacylation of 2-hydroxystyrenes. This work highlights the use of hydroacylation as a unified approach to both aldol and homoaldol products.
Product Description
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%