Merck
  • Determination of Co(II) in plant tissue by microwave digestion and ion chromatography coupled with luminol/perborate or luminol/percarbonate chemiluminescence detection.
Determination of Co(II) in plant tissue by microwave digestion and ion chromatography coupled with luminol/perborate or luminol/percarbonate chemiluminescence detection.
Phytochemical analysis : PCA (2010-09-08)
José A Murillo Pulgarín, Luisa F García Bermejo, Armando Carrasquero Durán
ABSTRACT
The cobalt is an essential element for leguminous plants but may be harmful for other species; for that reason determination of Co(II) is very important for the management of polluted areas and for discover plants with capacity for the hyperaccumulation of heavy metals, which has produced a growing necessity of fast, sensitive and selective analytical techniques. To develop an analytical procedure for the determination of cobalt in plant tissue by coupling the ionic chromatography to the luminol-based chemiluminescence detection. The sample was digested in a mixture of concentrated nitric acid and hydrogen peroxide, using an microwave oven to dissolve the Co(II). The solution containing Co(II) ions was injected to an ionic chromatograph using oxalic acid as the eluent. The detection was based on the catalytic effect of Co(II) on the luminol chemiluminescence using perborate or percarbonate as oxidants. Experimental variables, such as concentrations, pH, flow rates and acid digestion conditions were optimised. Well-resolved chromatographic peaks were obtained. The height and area showed linear dependences with the Co(II) concentration, which were used to quantify the heavy metal, with recoveries up to 95%. The microwave irradiation (60  s) was sufficient for the complete mineralisation of 200  mg of sample, employing 2  mL of the acid mixture. The method was free from the interferences, requiring less than 12 minutes to complete the analysis. The method was simple and rapid for the determination of cobalt in plant tissue with detection limits comparable to those obtained with more sophisticated and expensive analytical equipment.
Sigma-Aldrich
Sodium perborate monohydrate, powder, 20-100 mesh
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium perborate tetrahydrate, purum p.a., 96% (RT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium perborate tetrahydrate, 9-11% available Oxygen