Merck
  • Random mutants of a Pleurotus ostreatus laccase as new biocatalysts for industrial effluents bioremediation.
Random mutants of a Pleurotus ostreatus laccase as new biocatalysts for industrial effluents bioremediation.
Journal of applied microbiology (2009-09-09)
A Miele, P Giardina, G Sannia, V Faraco
ABSTRACT
To select better performing laccase variants among the 2300 randomly mutated variants of Pleurotus ostreatus POXA1b laccase to develop improved laccase-based biocatalysts. Screening of collections of 2300 randomly mutated variants of POXA1b was performed by assaying activity towards the phenolic substrate 2,6-dimethoxyphenol. Two new variants endowed with higher enzyme activity than the wild-type laccase were characterized, and their ability to decolourize industrial dyes with complex trisazo-, polyazo- and stilbene-type structures, in the absence of mediators, was demonstrated. One of the mutants (2L4A) was also proved to be highly stable at both acidic and alkaline pH values (displaying a half-life of around 1 month at the pH levels of both 5 and 10). In comparison with the wild-type laccase, the new selected 2L4A mutant shows a significant increase in stability at acidic pH, whilst storing its high stability at alkaline pH. This variant also represents a more versatile enzyme with respect to both the variety of xenobiotics degraded and the operative conditions. This work represents the first example of improvement of a basidiomycete laccase for industrial effluents bioremediation by directed evolution.
Sigma-Aldrich
2,6-Dimethoxyphenol, natural, ≥96%
Sigma-Aldrich
2,6-Dimethoxyphenol, 99%
Sigma-Aldrich
2,6-Dimethoxyphenol, ≥98%, FG
Supelco
2,6-Dimethoxyphenol, analytical standard