Merck
  • Crystallographic characterization of helical secondary structures in 2:1 and 1:2 alpha/beta-peptides.
Crystallographic characterization of helical secondary structures in 2:1 and 1:2 alpha/beta-peptides.
Journal of the American Chemical Society (2009-02-11)
Soo Hyuk Choi, Ilia A Guzei, Lara C Spencer, Samuel H Gellman
ABSTRACT
Oligomers containing both alpha- and beta-amino acid residues ("alpha/beta-peptides") are intriguing as potential foldamers. A large set of alpha/beta-peptide backbones can be generated by combining alpha- and beta-amino acid residues in different patterns; however, most research to date has focused on the simplest pattern, 1:1 alpha:beta. We have begun to explore the range of variation that can be achieved with alpha-residue/beta-residue combinations by examining the folding behavior of oligomers that contain 2:1 and 1:2 alpha:beta patterns. The beta-residues in our systems have a five-membered-ring constraint (trans-2-aminocyclopentanecarboxylic acid (ACPC) residues), because these preorganized subunits strongly promote helical folding for 1:1 alpha:beta backbones and pure beta backbones. Previously we concluded that two helical conformations are available to 2:1 and 1:2 alpha/beta-peptides containing ACPC or analogously constrained beta-residues, one helix defined by i,i+3 CO...H-N backbone hydrogen bonds and the other defined by i,i+4 CO...H-N hydrogen bonds. These deductions were based on 2D NMR analysis of a 2:1 heptamer and a 1:2 hexamer in methanol. Crystallographic analysis of a pair of analogous nonpolar alpha/beta-peptides showed only the i,i+3 hydrogen-bonded helical conformations. We now report four new crystal structures of 2:1 alpha/beta-peptides, ranging in length from 5 to 11 residues, and six new crystal structures of 1:2 alpha/beta-peptides, ranging in length from 6 to 10 residues. All 10 of these new structures are fully helical, and all helices display the i,i+3 CO...H-N hydrogen bonding pattern. These crystallographic data sets, collectively, provide high structural definition for the i,i+3 hydrogen-bonded helical secondary structures available to these foldamer backbones.
Sigma-Aldrich
Cycloleucine, 97%