Merck
  • Estimation of chromium(VI) sorption efficiency of novel regenerable p-tert-butylcalix[8]areneoctamide impregnated Amberlite resin.
Estimation of chromium(VI) sorption efficiency of novel regenerable p-tert-butylcalix[8]areneoctamide impregnated Amberlite resin.
Journal of hazardous materials (2008-10-03)
Imdadullah Qureshi, Shahabuddin Memon, Mustafa Yilmaz
ABSTRACT
The article describes a convenient synthesis and Cr(VI) extraction efficiency of a novel p-tert-butylcalix[8]areneoctamide impregnated Amberlite (XAD-4) resin. Using p-tert-butylcalix[8]arene macrocyclic building block, two strategies have been developed; i.e., derivatization of p-tert-butylcalix[8]arene framework with sophisticated ionophoric groups having efficiency to extract oxoanions from aqueous media and, impregnation of p-tert-butylcalix[8]arene derivative onto the polymeric support. Liquid-liquid and solid-liquid extraction experiments have been performed to evaluate the Cr(VI) extraction efficiency of both p-tert-butylcalix[8]arene derivative and the impregnated resin. From the results, it has been deduced that solid-phase extraction method is more convenient and efficient than liquid-liquid extraction process. The impregnated XAD-4 resin has been found regenerable with better extracting efficiency as compared to the simple p-tert-butylcalix[8]areneoctamide ionophore.
Product Description
Supelco
Amberlite XAD4, 20-60 mesh
Supelco
Amberlite XAD4, 20-60 mesh
Supelco
Amberlite XAD4
Supelco
Amberlite XAD4, 20-60 mesh