Merck
  • Birefringence dispersion of N-(4-methoxybenzylidene)-4-butylaniline (MBBA) determined from channeled spectra.
Birefringence dispersion of N-(4-methoxybenzylidene)-4-butylaniline (MBBA) determined from channeled spectra.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-02-20)
I Diaconu, Nicoleta Melniciuc-Puică, Dana Dorohoi, Magda Aflori
ABSTRACT
The birefringence of MBBA, determined from the channeled spectra, decreases when the wavelength increases, showing a normal dispersion. The changes induced by an external electrostatic field acting perpendicular on the nematic director in the birefringence values were evidenced. The birefringence increases with the electrostatic field intensity which enhances the preferential alignment by the big values of the electric dipole moments induced parallel to the long MBBA molecule. The main refractive indices were interferometrically measured for three visible monochromatic radiations and the birefringence values were concordant with those estimated from the channeled spectra. The main molecular polarizabilities were estimated.
Sigma-Aldrich
N-(4-Methoxybenzylidene)-4-butylaniline, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
0.2 mL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥98,300
100 μL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥98,300
0.5 mL
1 Available to ship on January 06, 2025
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1312
¥195,000
To order products, please contact your local dealer.