Merck
  • Buoyant density of DNA-Hoechst 33258 (bisbenzimide) complexes in CsCl gradients: Hoechst 33258 binds to single AT base pairs.
Buoyant density of DNA-Hoechst 33258 (bisbenzimide) complexes in CsCl gradients: Hoechst 33258 binds to single AT base pairs.
Analytical biochemistry (1991-04-01)
P Karlovsky, A W de Cock
ABSTRACT
Buoyant density of DNA in CsCl gradients with Hoechst 33258 (bisbenzimide) was investigated as a function of guanine plus cytosine content of the DNA (%GC; in mole percent). A formula for calculating %GC from the refractive index (nD) of the isopycnic CsCl/Hoechst 33258 solution over the range of 0-75 %GC was established: %GC = 351762.28 X nD - 123778.66 X nD2 - 249789.47 (the coefficients must not be rounded off). The shape of this curve indicates that under these conditions, in contrast to dilute buffers, Hoechst 33258 binds to single AT base pairs on DNA. Resolution of DNA bands in CsCl/Hoechst 33258 gradients is 1.6 to 2.1 times better than comparative CsCl gradients without the dye. Potential application to %GC determination is discussed.
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.