Merck
In vivo micronucleus test in mice with 1-phenylethanol.
Archives of toxicology (2006-09-01)
Günter Engelhardt
ABSTRACT
1-Phenylethanol is one of the major primary phase-I metabolites of ethylbenzene. In principle it may yield an electrophilic intermediate by phase-II metabolism. Because of the extensive use of ethylbenzene as a solvent, 2-year carcinogenicity inhalation studies were carried out leading to renal hyperplasia and tubular neoplasms both in male and female rats and alveolar/bronchiolar neoplasms in male mice and hepatocellular neoplasms in female mice. Whereas the mechanism underlying the increased renal tumor incidences in rats has been clarified, the mechanism of tumor formation (genotoxic or nongenotoxic mode of action) in the lung and liver of mice is still unclear. The genotoxicity data available to date for 1-phenylethanol include in vitro studies using either bacteria (Salmonella reverse mutation assay, E. coli Pol A(+)/Pol A(-) test) or mammalian cells (mouse lymphoma assay, chromosome aberration test and sister chromatid exchanges using CHO cells). These experiments, however, did not always follow current standard procedures and some of the data obtained are compromised and not always convincing. The present database thus does not allow a definitive assessment of the in vitro genotoxic potential of 1-phenylethanol. The in vitro database suggests that clastogenicity may be the most relevant genetic end point, and therefore an in vivo micronucleus assay in mouse bone marrow was carried out. The animals were given 1-phenylethanol in single oral doses up to the maximum tolerated dose of 750 mg/kg body weight. Bone marrow was sampled 24 and 48 h after treatment. Under the experimental conditions used, there was no evidence of increased micronuclei frequencies at any dose or sampling time. These findings indicate that 1-phenylethanol is not clastogenic in vivo. This information, together with other negative or inconclusive genotoxicity data available so far, suggests a nongenotoxic mode of action responsible for the lung and liver tumors observed in mice following 2 years of inhalation exposure to ethylbenzene.
Sigma-Aldrich
α-Methylbenzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
1-Phenylethanol, 98%