Merck
  • Aerobic cometabolism of chloroform by butane-grown microorganisms: long-term monitoring of depletion rates and isolation of a high-performing strain.
Aerobic cometabolism of chloroform by butane-grown microorganisms: long-term monitoring of depletion rates and isolation of a high-performing strain.
Biodegradation (2005-02-26)
Dario Frascari, Arianna Zannoni, Stefano Fedi, Youry Pii, Davide Zannoni, Davide Pinelli, Massimo Nocentini
ABSTRACT
The focus of this microcosm study was to monitor the performances of 17 butane-utilizing microcosms during a long-term (100-250 days) aerobic cometabolic depletion of chloroform (CF). The depletion of the contaminant began after a lag-time variable between 0 and 23 days. All microcosms quickly reached a pseudo steady-state condition, in terms of biomass concentration (with an average of 9.3 x 106 CFU ml(-1)), chloroform depletion rate (5 micromol l(-1) d(-1)) and butane utilization rate (730 micromol l(-1) d(-1)). After about 100 days of CF depletion, a sudden 5- to 7-fold increase of the chloroform rate was observed in two microcosms, where the highest amount of contaminant had been depleted. In one of these high-performing microcosms, an experiment of chloroform depletion in the absence of butane resulted in the depletion of a surprisingly high amount of contaminant (765 micromolCF kg(-1) dry soil in 2 months) and in a marked selection of a single bacterial strain. Bioaugmentation assays conducted with the biomass selected in this microcosm and with a pure culture of the selected strain immediately resulted in very high chloroform depletion rates. Preliminary results of a study conducted with resting cells of the selected strain indicated that it can degrade chloroform concentrations up to 119 microM (14.2 mg l(-1)) without any sign of substrate toxicity, and that it is able to transform vinyl chloride and 1,1,2-trichloroethane.
Product Description
Supelco
1,1,2-Trichloroethane, analytical standard
Sigma-Aldrich
1,1,2-Trichloroethane, contains ≤3% 2-propanol as stabilizer, 97%