Merck
Probing every layer in dendrons.
Journal of the American Chemical Society (2005-02-17)
Kulandaivelu Sivanandan, Sivakumar V Aathimanikandan, Christopher G Arges, Christopher J Bardeen, S Thayumanavan
ABSTRACT
Dendrons with a fluorescent probe in a specific location have been synthesized and characterized. Accessibility of guest molecules to each of these layers was then analyzed using an intermolecular photoinduced electron-transfer process. Comparisons of the accessibility among each layer of the dendron and among the generations are provided.
Sigma-Aldrich
Benzyl ether, ≥98%, FCC, FG
Sigma-Aldrich
Dibenzyl ether, purum, ≥98.0% (GC)
Sigma-Aldrich
Benzyl ether, 98%