Merck
  • Synthesis of beta-lactones by the regioselective, cobalt and Lewis acid catalyzed carbonylation of simple and functionalized epoxides.
Synthesis of beta-lactones by the regioselective, cobalt and Lewis acid catalyzed carbonylation of simple and functionalized epoxides.
The Journal of organic chemistry (2001-08-04)
J T Lee, P J Thomas, H Alper
ABSTRACT
The PPNCo(CO)(4) and BF(3) x Et(2)O catalyzed carbonylation of simple and functionalized epoxides in DME gives the corresponding beta-lactones regioselectively in good to high yields. The carbonylation occurred selectively at the unsubstituted C-O bond of the epoxide ring, and this reaction tolerates various functional groups such as alkenyl, halide, hydroxy, and alkyl ether.
Sigma-Aldrich
β-Butyrolactone, 98%