Merck
  • Catalytic and molecular properties of the quinohemoprotein tetrahydrofurfuryl alcohol dehydrogenase from Ralstonia eutropha strain Bo.
Catalytic and molecular properties of the quinohemoprotein tetrahydrofurfuryl alcohol dehydrogenase from Ralstonia eutropha strain Bo.
Journal of bacteriology (2001-02-27)
G Zarnt, T Schräder, J R Andreesen
ABSTRACT
The quinohemoprotein tetrahydrofurfuryl alcohol dehydrogenase (THFA-DH) from Ralstonia eutropha strain Bo was investigated for its catalytic properties. The apparent k(cat)/K(m) and K(i) values for several substrates were determined using ferricyanide as an artificial electron acceptor. The highest catalytic efficiency was obtained with n-pentanol exhibiting a k(cat)/K(m) value of 788 x 10(4) M(-1) s(-1). The enzyme showed substrate inhibition kinetics for most of the alcohols and aldehydes investigated. A stereoselective oxidation of chiral alcohols with a varying enantiomeric preference was observed. Initial rate studies using ethanol and acetaldehyde as substrates revealed that a ping-pong mechanism can be assumed for in vitro catalysis of THFA-DH. The gene encoding THFA-DH from R. eutropha strain Bo (tfaA) has been cloned and sequenced. The derived amino acid sequence showed an identity of up to 67% to the sequence of various quinoprotein and quinohemoprotein dehydrogenases. A comparison of the deduced sequence with the N-terminal amino acid sequence previously determined by Edman degradation analysis suggested the presence of a signal sequence of 27 residues. The primary structure of TfaA indicated that the protein has a tertiary structure quite similar to those of other quinoprotein dehydrogenases.
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, ≥98%
Pricing and availability is not currently available.