Merck
  • Human NK cells kill resting but not activated microglia via NKG2D- and NKp46-mediated recognition.
Human NK cells kill resting but not activated microglia via NKG2D- and NKp46-mediated recognition.
Journal of immunology (Baltimore, Md. : 1950) (2008-10-23)
Anna Lünemann, Jan D Lünemann, Susanne Roberts, Brady Messmer, Rosa Barreira da Silva, Cedric S Raine, Christian Münz
ABSTRACT
Microglia are resident macrophage-like APCs of the CNS. To avoid escalation of inflammatory processes and bystander damage within the CNS, microglia-driven inflammatory responses need to be tightly regulated and both spatially and temporally restricted. Following traumatic, infectious, and autoimmune-mediated brain injury, NK cells have been found in the CNS, but the functional significance of NK cell recruitment and their mechanisms of action during brain inflammation are not well understood. In this study, we investigated whether and by which mechanisms human NK cells might edit resting and activated human microglial cells via killing in vitro. IL-2-activated NK cells efficiently killed both resting allogeneic and autologous microglia in a cell-contact-dependent manner. Activated NK cells rapidly formed synapses with human microglial cells in which perforin had been polarized to the cellular interface. Ab-mediated NKG2D and NKp46 blockade completely prevented the killing of human microglia by activated NK cells. Up-regulation of MHC class I surface expression by TLR4 stimulation protected microglia from NK cell-mediated killing, whereas MHC class I blockade enhanced cytotoxic NK cell activity. These data suggest that brain-infiltrating NK cells might restrict innate and adaptive immune responses within the human CNS via elimination of resting microglia.
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Sigma-Aldrich
Saponin, for molecular biology, used as non-ionic surfactant
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by phenol extraction