Merck
  • Establishment of an Electrophysiological Platform for Modeling ALS with Regionally-Specific Human Pluripotent Stem Cell-Derived Astrocytes and Neurons.
Establishment of an Electrophysiological Platform for Modeling ALS with Regionally-Specific Human Pluripotent Stem Cell-Derived Astrocytes and Neurons.
Journal of visualized experiments : JoVE (2021-09-14)
Arens Taga, Christa W Habela, Alexandra Johns, Shiyu Liu, Mollie O'Brien, Nicholas J Maragakis
ABSTRACT
Human pluripotent stem cell-derived astrocytes (hiPSC-A) and neurons (hiPSC-N) provide a powerful tool for modeling Amyotrophic Lateral Sclerosis (ALS) pathophysiology in vitro. Multi-electrode array (MEA) recordings are a means to record electrical field potentials from large populations of neurons and analyze network activity over time. It was previously demonstrated that the presence of hiPSC-A that are differentiated using techniques to promote a spinal cord astrocyte phenotype improved maturation and electrophysiological activity of regionally specific spinal cord hiPSC-motor neurons (MN) when compared to those cultured without hiPSC-A or in the presence of rodent astrocytes. Described here is a method to co-culture spinal cord hiPSC-A with hiPSC-MN and record electrophysiological activity using MEA recordings. While the differentiation protocols described here are particular to astrocytes and neurons that are regionally specific to the spinal cord, the co-culturing platform can be applied to astrocytes and neurons differentiated with techniques specific to other fates, including cortical hiPSC-A and hiPSC-N. These protocols aim to provide an electrophysiological assay to inform about glia-neuron interactions and provide a platform for testing drugs with therapeutic potential in ALS.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
Purmorphamine, InSolution, ≥98%, 50 mM in DMSO, induces osteoblast differentiation in C3H10T1/2 multipotent mesenchymal progenitor cells