Merck
  • Colesevelam ameliorates non-alcoholic steatohepatitis and obesity in mice.
Colesevelam ameliorates non-alcoholic steatohepatitis and obesity in mice.
Hepatology international (2022-01-26)
Phillipp Hartmann, Yi Duan, Yukiko Miyamoto, Münevver Demir, Sonja Lang, Elda Hasa, Patrick Stern, Dennis Yamashita, Mary Conrad, Lars Eckmann, Bernd Schnabl
ABSTRACT
Obesity, non-alcoholic fatty liver disease (NAFLD) and its more advanced form non-alcoholic steatohepatitis (NASH) are important causes of morbidity and mortality worldwide. Bile acid dysregulation is a pivotal part in their pathogenesis. The aim of this study was to evaluate the bile acid sequestrant colesevelam in a microbiome-humanized mouse model of diet-induced obesity and steatohepatitis. Germ-free C57BL/6 mice were associated with stool from patients with NASH and subjected to 20 weeks of Western diet feeding with and without colesevelam. Colesevelam reduced Western diet-induced body and liver weight gain in microbiome-humanized mice compared with controls. It ameliorated Western diet-induced hepatic inflammation, steatosis, fibrosis and insulin resistance. Colesevelam increased de novo bile acid synthesis and decreased hepatic cholesterol content in microbiome-humanized mice fed a Western diet. It further induced the gene expression of the antimicrobials Reg3g and Reg3b in the distal small intestine and decreased plasma levels of LPS. Colesevelam ameliorates Western diet-induced steatohepatitis and obesity in microbiome-humanized mice.
Sigma-Aldrich
Colesevelam hydrochloride, ≥98% (HPLC)