Merck
TNFα- and IL-17A-mediated S100A8 expression is regulated by p38 MAPK.
Experimental dermatology (2013-06-27)
Maike Mose, Zhanyuan Kang, Line Raaby, Lars Iversen, Claus Johansen
ABSTRACT
The antimicrobial peptide S100A8 is known to be upregulated in lesional psoriatic skin compared with non-lesional psoriatic skin and is believed to play a role in the pathogenesis of psoriasis. However, little is known about the signalling pathways involved in the regulation of S100A8 expression. Using quantitative real-time RT-PCR analysis, we demonstrated that stimulation with TNFα and IL-17A in combination resulted in a significant and synergistic induction of S100A8 mRNA in human keratinocytes. TNFα and IL-17A also induced the S100A8 promoter activity synergistically. This was demonstrated by a gene reporter assay in cells transfected with a luciferase plasmid construct, consisting of 3502 base pairs of the human S100A8 promoter. The TNFα- and IL-17A-mediated induction of S100A8 mRNA and protein was mediated by a p38 MAPK-dependent mechanism, as demonstrated by the use of a p38 MAPK inhibitor. Finally, adalimumab treatment for patients with psoriasis significantly decreased S100A8 mRNA at day fourteen after start of treatment, but not at day four. Taken together, this study demonstrates that the p38 MAPK signalling pathway plays a key role in the TNFα- and IL-17A-induced expression of S100A8 in cultured human keratinocytes.
Sigma-Aldrich
IKK-2 Inhibitor, SC-514, The IKK-2 Inhibitor, SC-514, also referenced under CAS 354812-17-2, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Millipore
SB 202190, SB 202190, CAS 152121-30-7, is a potent, reversible, competitive inhibitor of p38. Inhibits p38 phosphorylation of myelin basic protein. Blocks the activity of p38β (Ki = 16 nM; IC₅₀ = 350 nM).
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.