- Profiling DNA methylome landscapes of mammalian cells with single-cell reduced-representation bisulfite sequencing.
Profiling DNA methylome landscapes of mammalian cells with single-cell reduced-representation bisulfite sequencing.
The heterogeneity of DNA methylation within a population of cells necessitates DNA methylome profiling at single-cell resolution. Recently, we developed a single-cell reduced-representation bisulfite sequencing (scRRBS) technique in which we modified the original RRBS method by integrating all the experimental steps before PCR amplification into a single-tube reaction. These modifications enable scRRBS to provide digitized methylation information on ā¼1 million CpG sites within an individual diploid mouse or human cell at single-base resolution. Compared with the single-cell bisulfite sequencing (scBS) technique, scRRBS covers fewer CpG sites, but it provides better coverage for CpG islands (CGIs), which are likely to be the most informative elements for DNA methylation. The entire procedure takes ā¼3 weeks, and it requires strong molecular biology skills.