Merck
  • Functional Analysis of Human and Feline Coronavirus Cross-Reactive Antibodies Directed Against the SARS-CoV-2 Fusion Peptide.
Functional Analysis of Human and Feline Coronavirus Cross-Reactive Antibodies Directed Against the SARS-CoV-2 Fusion Peptide.
Frontiers in immunology (2022-01-25)
Nathalie Vanderheijden, Annelies Stevaert, Jiexiong Xie, Xiaolei Ren, Cyril Barbezange, Sam Noppen, Isabelle Desombere, Bruno Verhasselt, Peter Geldhof, Nick Vereecke, Veerle Stroobants, Dayoung Oh, Merijn Vanhee, Lieve M J Naesens, Hans J Nauwynck
ABSTRACT
To face the continuous emergence of SARS-CoV-2 variants, broadly protective therapeutic antibodies are highly needed. We here focused on the fusion peptide (FP) region of the viral spike antigen since it is highly conserved among alpha- and betacoronaviruses. First, we found that coronavirus cross-reactive antibodies are commonly formed during infection, being omnipresent in sera from COVID-19 patients, in ~50% of pre-pandemic human sera (rich in antibodies against endemic human coronaviruses), and even in feline coronavirus-infected cats. Pepscan analyses demonstrated that a confined N-terminal region of the FP is strongly immunogenic across diverse coronaviruses. Peptide-purified human antibodies targeting this conserved FP epitope exhibited broad binding of alpha- and betacoronaviruses, besides weak and transient SARS-CoV-2 neutralizing activity. Being frequently elicited by coronavirus infection, these FP-binding antibodies might potentially exhibit Fc-mediated effector functions and influence the kinetics or severity of coronavirus infection and disease.
Sigma-Aldrich
Anti-Coronavirus Group Antigen Antibody, nucleoprotein of OC-43, clone 542-7D, clone 542-7D, Chemicon®, from mouse
Roche
Neuraminidase (Sialidase), from Vibrio cholerae