Merck
  • AHL signals induce rubrifacine production in a bruI mutant of Brenneria rubrifaciens.
AHL signals induce rubrifacine production in a bruI mutant of Brenneria rubrifaciens.
Phytopathology (2012-01-13)
Ali E McClean, Breck A Duerkop, E Peter Greenberg, Daniel A Kluepfel
ABSTRACT
Several members of the bacterial genus Brenneria are pathogenic on different tree species. Cell-free extracts from the bacterial phytopathogens Brenneria rubrifaciens, B. salicis, and B. nigrifluens induced production of the red pigment rubrifacine in the B. rubrifaciens bruI insertional mutant Br-212. Analysis of the bruI locus identified an adjacent open reading frame, designated bruR, with homology to luxR. High-performance liquid chromatography and mass spectrometry analysis of ethyl acetate extracts from wild-type B. rubrifaciens and Escherichia coli expressing the bruI gene identified two acyl homoserine lactone (AHL) peaks, N-(3-oxohexanoyl)-homoserine lactone (3OC6HSL) and N-hexanoyl-homoserine lactone (C6HSL). Addition of synthetic 3OC6HSL and C6HSL at 10 μM to the bruI mutant, strain Br-212, induced rubrifacine production and the ability to elicit a hypersensitive reaction (HR) in tobacco leaves. Synthetic C6HSL was less effective at inducing pigment production than 3OC6HSL at 10 μM. The bruI mutant Br-212 did not produce detectable AHLs, indicating that C6HSL and 3OC6HSL are the major AHLs produced by this species. The AHLs N-heptanoyl-DL-homoserine lactone (C7HSL), N-octanoyl-DL-homoserine lactone (C8HSL), and N-(3-oxooctanoyl)-DL-homoserine lactone (3OC8HSL) also induced pigment production in Br-212 and restored its ability to elicit an HR in tobacco, suggesting that cross-talk with other bacterial species may be possible.
Sigma-Aldrich
N-Heptanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Octanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Tetradecanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Sigma-Aldrich
N-Dodecanoyl-DL-homoserine lactone, ≥97.0% (HPLC)