Merck
Oxidation of amines by flavoproteins.
Archives of biochemistry and biophysics (2009-08-05)
Paul F Fitzpatrick
ABSTRACT
Many flavoproteins catalyze the oxidation of primary and secondary amines, with the transfer of a hydride equivalent from a carbon-nitrogen bond to the flavin cofactor. Most of these amine oxidases can be classified into two structural families, the D-amino acid oxidase/sarcosine oxidase family and the monoamine oxidase family. This review discusses the present understanding of the mechanisms of amine and amino acid oxidation by flavoproteins, focusing on these two structural families.
Sigma-Aldrich
Sarcosine Oxidase from Bacillus sp., lyophilized powder, 25-50 units/mg solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
1 g
1 Estimated to ship on January 27, 2025
Details...
€63.60
5 g
1 Estimated to ship on January 27, 2025
Details...
€231.00
10 g
1 Estimated to ship on February 24, 2025
Details...
€390.00
25 g
1 Estimated to ship on February 24, 2025
Details...
€778.00
To order products, please contact your local dealer.