Merck
  • Emerging lysophospholipid mediators, lysophosphatidylserine, lysophosphatidylthreonine, lysophosphatidylethanolamine and lysophosphatidylglycerol.
Emerging lysophospholipid mediators, lysophosphatidylserine, lysophosphatidylthreonine, lysophosphatidylethanolamine and lysophosphatidylglycerol.
Prostaglandins & other lipid mediators (2009-05-12)
Kumiko Makide, Hajime Kitamura, Yusuke Sato, Michiyo Okutani, Junken Aoki
ABSTRACT
It is now widely accepted that lysophospholipids (LPLs), a product of the phospholipase A reaction, function as mediators through G-protein-coupled receptors. Notably, recent studies of lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P) have revealed their essential roles in vivo. On the other hand, other LPLs such as lysophosphatidylserine (LPS), lysophosphatidylthreonine (LPT), lysophosphatidylethanolamine (LPE), lysophosphatidylinositol (LPI) and lysophosphatidylglycerol (LPG) have been reported to show lipid mediator-like responses both in vivo (LPS and LPT) and in vitro (LPS, LPT, LPE and LPG), while very little is known about their receptor, synthetic enzyme and patho-physiological roles. In this review, we summarize the actions of these LPLs as lipid mediators including LPS, LPT, LPE and LPG.
Product Description
Avanti
18:1-d7 Lyso PE, Avanti Research - A Croda Brand 791644C
Sigma-Aldrich
3-sn-Lysophosphatidylethanolamine from egg yolk, Type I, ≥99%