Merck
  • Polypurine-repeat-containing RNAs: a novel class of long non-coding RNA in mammalian cells.
Polypurine-repeat-containing RNAs: a novel class of long non-coding RNA in mammalian cells.
Journal of cell science (2010-10-14)
Ruiping Zheng, Zhen Shen, Vidisha Tripathi, Zhenyu Xuan, Susan M Freier, C Frank Bennett, Supriya G Prasanth, Kannanganattu V Prasanth
ABSTRACT
In higher eukaryotic cells, long non-protein-coding RNAs (lncRNAs) have been implicated in a wide array of cellular functions. Cell- or tissue-specific expression of lncRNA genes encoded in the mammalian genome is thought to contribute to the complex gene networks needed to regulate cellular function. Here, we have identified a novel species of polypurine triplet repeat-rich lncRNAs, designated as GAA repeat-containing RNAs (GRC-RNAs), that localize to numerous punctate foci in the mammalian interphase nuclei. GRC-RNAs consist of a heterogeneous population of RNAs, ranging in size from ~1.5 kb to ~4 kb and localize to subnuclear domains, several of which associate with GAA.TTC-repeat-containing genomic regions. GRC-RNAs are components of the nuclear matrix and interact with various nuclear matrix-associated proteins. In mitotic cells, GRC-RNAs form distinct cytoplasmic foci and, in telophase and G1 cells, localize to the midbody, a structure involved in accurate cell division. Differentiation of tissue culture cells leads to a decrease in the number of GRC-RNA nuclear foci, albeit with an increase in size as compared with proliferating cells. Conversely, the number of GRC-RNA foci increases during cellular transformation. We propose that nuclear GRC-RNAs represent a novel family of mammalian lncRNAs that might play crucial roles in the cell nucleus.
Sigma-Aldrich
Anti-hnRNP Q Antibody, clone 18E4, clone 18E4, from mouse
Sigma-Aldrich
Anti-Histidine Tagged Antibody, clone 4D11, clone 4D11, Upstate®, from mouse