Merck
  • Proteomic Analysis Uncovers Measles Virus Protein C Interaction With p65-iASPP Protein Complex.
Proteomic Analysis Uncovers Measles Virus Protein C Interaction With p65-iASPP Protein Complex.
Molecular & cellular proteomics : MCP (2021-01-31)
Alice Meignié, Chantal Combredet, Marc Santolini, István A Kovács, Thibaut Douché, Quentin Giai Gianetto, Hyeju Eun, Mariette Matondo, Yves Jacob, Regis Grailhe, Frédéric Tangy, Anastassia V Komarova
ABSTRACT
Viruses manipulate the central machineries of host cells to their advantage. They prevent host cell antiviral responses to create a favorable environment for their survival and propagation. Measles virus (MV) encodes two nonstructural proteins MV-V and MV-C known to counteract the host interferon response and to regulate cell death pathways. Several molecular mechanisms underlining MV-V regulation of innate immunity and cell death pathways have been proposed, whereas MV-C host-interacting proteins are less studied. We suggest that some cellular factors that are controlled by MV-C protein during viral replication could be components of innate immunity and the cell death pathways. To determine which host factors are targeted by MV-C, we captured both direct and indirect host-interacting proteins of MV-C protein. For this, we used a strategy based on recombinant viruses expressing tagged viral proteins followed by affinity purification and a bottom-up mass spectrometry analysis. From the list of host proteins specifically interacting with MV-C protein in different cell lines, we selected the host targets that belong to immunity and cell death pathways for further validation. Direct protein interaction partners of MV-C were determined by applying protein complementation assay and the bioluminescence resonance energy transfer approach. As a result, we found that MV-C protein specifically interacts with p65-iASPP protein complex that controls both cell death and innate immunity pathways and evaluated the significance of these host factors on virus replication.
Product Description
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Supelco
p-Iodoclonidine hydrochloride, analytical standard, for drug analysis
Sigma-Aldrich
Anti-Measles Antibody, nucleoprotein, clone 83KKII, FITC-conjugated, clone 83KKII, Chemicon®, from mouse