Merck
  • SPF45/RBM17-dependent, but not U2AF-dependent, splicing in a distinct subset of human short introns.
SPF45/RBM17-dependent, but not U2AF-dependent, splicing in a distinct subset of human short introns.
Nature communications (2021-08-15)
Kazuhiro Fukumura, Rei Yoshimoto, Luca Sperotto, Hyun-Seo Kang, Tetsuro Hirose, Kunio Inoue, Michael Sattler, Akila Mayeda
ABSTRACT
Human pre-mRNA introns vary in size from under fifty to over a million nucleotides. We searched for essential factors involved in the splicing of human short introns by screening siRNAs against 154 human nuclear proteins. The splicing activity was assayed with a model HNRNPH1 pre-mRNA containing short 56-nucleotide intron. We identify a known alternative splicing regulator SPF45 (RBM17) as a constitutive splicing factor that is required to splice out this 56-nt intron. Whole-transcriptome sequencing of SPF45-deficient cells reveals that SPF45 is essential in the efficient splicing of many short introns. To initiate the spliceosome assembly on a short intron with the truncated poly-pyrimidine tract, the U2AF-homology motif (UHM) of SPF45 competes out that of U2AF65 (U2AF2) for binding to the UHM-ligand motif (ULM) of the U2 snRNP protein SF3b155 (SF3B1). We propose that splicing in a distinct subset of human short introns depends on SPF45 but not U2AF heterodimer.
Sigma-Aldrich
Anti-SF4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RBM17 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-U2AF65 antibody, Mouse monoclonal, clone MC3, purified from hybridoma cell culture