Merck
  • A splicing variant of TFEB negatively regulates the TFEB-autophagy pathway.
A splicing variant of TFEB negatively regulates the TFEB-autophagy pathway.
Scientific reports (2021-10-28)
Jee-Yun Park, Hee-Young Sohn, Young Ho Koh, Chulman Jo
ABSTRACT
Transcription factor EB (TFEB) is a master regulator of the autophagy-lysosomal pathway (ALP). Here, we cloned a novel splicing variant of TFEB, comprising 281 amino acids (hereafter referred to as small TFEB), and lacking the helix-loop-helix (HLH) and leucine zipper (LZ) motifs present in the full-length TFEB (TFEB-L). The TFEB variant is widely expressed in several tissues, including the brain, although its expression level is considerably lower than that of TFEB-L. Intriguingly, in cells stably expressing small TFEB, the expression profile of genes was inverted compared to that in cells ectopically expressing TFEB-L. In addition, fisetin-induced luciferase activity of promoter containing either coordinated lysosomal expression and regulation (CLEAR) element or antioxidant response element (ARE) was significantly repressed by co-transfection with small TFEB. Moreover, fisetin-mediated clearance of phosphorylated tau or α-synuclein was attenuated in the presence of small TFEB. Taken together, the results suggest that small TFEB is a novel splicing variant of TFEB that might act as a negative regulator of TFEB-L, thus fine tuning the activity of ALP during cellular stress.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
mTOR Inhibitor XI, Torin1, mTOR Inhibitor XI, Torin1, CAS 1222998-36-8, is a cell-permeable, highly potent, ATP-competitive inhibitor of mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively).
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)