Merck
  • Changes to the identity of EndoC-βH1 beta cells may be mediated by stress-induced depletion of HNRNPD.
Changes to the identity of EndoC-βH1 beta cells may be mediated by stress-induced depletion of HNRNPD.
Cell & bioscience (2021-07-25)
Nicola Jeffery, David Chambers, Brandon M Invergo, Ryan M Ames, Lorna W Harries
ABSTRACT
Beta cell identity changes occur in the islets of donors with diabetes, but the molecular basis of this remains unclear. Protecting residual functional beta cells from cell identity changes may be beneficial for patients with diabetes. A somatostatin-positive cell population was induced in stressed clonal human EndoC-βH1 beta cells and was isolated using FACS. A transcriptomic characterisation of somatostatin-positive cells was then carried out. Gain of somatostatin-positivity was associated with marked dysregulation of the non-coding genome. Very few coding genes were differentially expressed. Potential candidate effector genes were assessed by targeted gene knockdown. Targeted knockdown of the HNRNPD gene induced the emergence of a somatostatin-positive cell population in clonal EndoC-βH1 beta cells comparable with that we have previously reported in stressed cells. We report here a role for the HNRNPD gene in determination of beta cell identity in response to cellular stress. These findings widen our understanding of the role of RNA binding proteins and RNA biology in determining cell identity and may be important for protecting remaining beta cell reserve in diabetes.
Sigma-Aldrich
TRI Reagent®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
50 mg
1 Available to ship on December 26, 2024
Details...
$171.00
To order products, please contact your local dealer.