Merck
  • Histological and immunohistological investigation of lymphoid tissue in the platypus (Ornithorhynchus anatinus).
Histological and immunohistological investigation of lymphoid tissue in the platypus (Ornithorhynchus anatinus).
Journal of anatomy (1999-10-21)
J H Connolly, P J Canfield, S J McClure, R J Whittington
ABSTRACT
The gross and histological appearance and the distribution of T and B lymphocytes and plasma cells are described for lymphoid tissues obtained from 15 platypuses. The spleen was bilobed and surrounded by a thick capsule of collagen, elastic fibres and little smooth muscle. White pulp was prominent and included germinal centres and periarterial lymphoid sheaths. Red pulp contained haematopoietic tissue. A thin lobulated thymus was located within the mediastinum overlying the heart. The cortex of lobules consisted of dense aggregates of small and medium lymphocytes, scattered macrophages and few reticular epithelial cells. In the medulla, Hassall's corpuscles were numerous, lymphocytes were small and less abundant, and reticular cells were more abundant than in the cortex. Lymphoid nodules scattered throughout loose connective tissue in cervical, pharyngeal, thoracic, mesenteric and pelvic sites measured 790 +/- 370 microm (mean +/- S.D., n = 39) in diameter, the larger of which could be observed macroscopically. These consisted of single primary or secondary follicles supported by a framework of reticular fibres. Macrophages were common in the germinal centres. The platypus had a full range of gut-associated lymphoid tissue. No tonsils were observed macroscopically but histologically they consisted of submucosal follicles and intraepithelial lymphocytes. Peyer's patches were not observed macroscopically but histologically they consisted of several prominent submucosal secondary follicles in the antimesenteric wall of the intestine. Caecal lymphoid tissue consisted of numerous secondary follicles in the submucosa and densely packed lymphocytes in the lamina propria. Bronchus-associated lymphoid tissue was not observed macroscopically but was identified in 7 of 11 platypus lungs assessed histologically. Lymphoid cells were present as primary follicles associated with bronchi, as aggregates adjacent to blood vessels and as intraepithelial lymphocytes. The distribution of T lymphocytes, identified with antihuman CD3 and CD5, and B lymphocytes and plasma cells, identified with antihuman CD79a and CD79b and antiplatypus immunoglobulin, within lymphoid tissues in the platypus was similar to that described in therian mammals except for an apparent relative paucity of B lymphocytes. This study establishes that the platypus has a well-developed lymphoid system which is comparable in histological structure to that in therian mammals. It also confirms the distinctiveness of its peripheral lymphoid tissue, namely lymphoid nodules. Platypus lymphoid tissue has all the essential cell types, namely T and B lymphocytes and plasma cells, to mount an effective immune response against foreign antigens.
Sigma-Aldrich
HAT Media Supplement (50×) Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
100 mg
1 Available to ship on January 06, 2025
Details...
$92.20
To order products, please contact your local dealer.