Merck
  • Maklamicin, an antibacterial polyketide from an endophytic Micromonospora sp.
Maklamicin, an antibacterial polyketide from an endophytic Micromonospora sp.
Journal of natural products (2011-03-11)
Yasuhiro Igarashi, Hiromu Ogura, Kazuo Furihata, Naoya Oku, Chantra Indananda, Arinthip Thamchaipenet
ABSTRACT
A new spirotetronate-class polyketide, maklamicin (1), was isolated from the culture extract of an endophytic actinomycete of the genus Micromonospora. The structure and relative configuration of 1 were elucidated by interpretation of NMR and other spectroscopic data, and the absolute configuration was determined using the modified Mosher method. Maklamicin (1) showed strong to modest antimicrobial activity against Gram-positive bacteria.
Product Description
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)