Merck
  • Influence of Vanillin Acrylate-Based Resin Composition on Resin Photocuring Kinetics and Antimicrobial Properties of the Resulting Polymers.
Influence of Vanillin Acrylate-Based Resin Composition on Resin Photocuring Kinetics and Antimicrobial Properties of the Resulting Polymers.
Materials (Basel, Switzerland) (2021-02-13)
Aukse Navaruckiene, Danguole Bridziuviene, Vita Raudoniene, Egidija Rainosalo, Jolita Ostrauskaite
ABSTRACT
The investigation of the influence of vanillin acrylate-based resin composition on photocuring kinetics and antimicrobial properties of the resulting polymers was performed in order to find efficient photocurable systems for optical 3D printing of bio-based polymers with tunable rigidity, as well as with antibacterial and antifungal activity. Two vanillin derivatives, vanillin diacrylate and vanillin dimethacrylate, were tested in photocurable systems using phenyl bis(2,4,6-trimethylbenzoyl)phosphine oxide as a photoinitiator. The influence of vanillin acrylate monomer, amount of photoinitiator, presence and amount of dithiol, and presence of solvent on photocuring kinetics was investigated by real-time photoreometry. Polymers of different rigidity were obtained by changing the photocurable resin composition. The photocuring kinetics of the selected vanillin acrylate-based resins was comparable with that of commercial petroleum-based acrylate resins for optical 3D printing. Polymers based on both vanillin acrylates showed a significant antibacterial activity against Escherichia coli and Staphylococcus aureus. Vanillin diacrylate-based polymer films also demonstrated an antifungal activity in direct contact with Aspergillus niger and Aspergillus terreus. Vanillin diacrylate-based dual curing systems were selected as the most promising for optical 3D printing of bio-based polymers with antibacterial and antifungal activity.
Sigma-Aldrich
6-Mercaptohexanoic acid, 90%
Sigma-Aldrich
Biphenyl-4,4′-dithiol, 95%