Merck
  • BV-2 Microglial Cells Respond to Rotenone Toxic Insult by Modifying Pregnenolone, 5α-Dihydroprogesterone and Pregnanolone Levels.
BV-2 Microglial Cells Respond to Rotenone Toxic Insult by Modifying Pregnenolone, 5α-Dihydroprogesterone and Pregnanolone Levels.
Cells (2020-09-17)
Rossella Avallone, Chiara Lucchi, Giulia Puja, Alessandro Codeluppi, Monica Filaferro, Giovanni Vitale, Cecilia Rustichelli, Giuseppe Biagini
ABSTRACT
Neuroinflammation, whose distinctive sign is the activation of microglia, is supposed to play a key role in the development and progression of neurodegenerative diseases. The aim of this investigation was to determine levels of neurosteroids produced by resting and injured BV-2 microglial cells. BV-2 cells were exposed to increasing concentrations of rotenone to progressively reduce their viability by increasing reactive oxygen species (ROS) production. BV-2 cell viability was significantly reduced 24, 48 and 72 h after rotenone (50-1000 nM) exposure. Concomitantly, rotenone (50-100 nM) determined a dose-independent augmentation of ROS production. Then, BV-2 cells were exposed to a single, threshold dose of rotenone (75 nM) to evaluate the overtime release of neurosteroids. In particular, pregnenolone, pregnenolone sulfate, progesterone, 5α-dihydroprogesterone (5α-DHP), allopregnanolone, and pregnanolone, were quantified in the culture medium by liquid chromatography with tandem mass spectrometry (LC-MS/MS) analysis. BV-2 cells synthesized all the investigated neurosteroids and, after exposure to rotenone, 5αDHP and pregnanolone production was remarkably increased. In conclusion, we found that BV-2 cells not only synthesize several neurosteroids, but further increase this production following oxidative damage. Pregnanolone and 5α-DHP may play a role in modifying the progression of neuroinflammation in neurodegenerative diseases.
Sigma-Aldrich
Pregnenolone-20,21-13C2-16,16-d2 sulfate sodium salt, ≥98 atom %, 98% (CP)
Pricing and availability is not currently available.