Merck
  • MiR-1247-3p protects rat cardiomyocytes against hypoxia/reoxygenation-induced injury via targeting BCL2L11 and caspase-2.
MiR-1247-3p protects rat cardiomyocytes against hypoxia/reoxygenation-induced injury via targeting BCL2L11 and caspase-2.
Journal of receptor and signal transduction research (2020-07-02)
Jun Huang, Yiteng Huang, Zongming Feng, Wenyu Guo, Xiaoqing Wang, Zhiyong Liao
ABSTRACT
Acute myocardial infarction (AMI) represents a severe coronary heart disease with relatively high rate of mortality and usually can lead to the damage of the myocardial tissues. Reperfusion of the ischemic myocardial tissues can minimize AMI-induced damage. As far as we know, the molecular mechanisms underlying ischemia/reperfusion (I/R)-induced injury remains elusive. This study was undertaken to explore the role of miR-1247-3p in regulating myocardial I/R injury. The hypoxia/reoxygenation (H/R)-treated H9c2 cells showed a decreased cell viability and mitochondrial membrane potential with an increase in the apoptosis; furthermore, miR-1247-3p was down-regulated in these cells. MiR-1247-3p overexpression attenuated H/R-induced H9c2 cell injury; while miR-1247-3p knockdown in H9c2 cells exhibited similar effects being observed in H/R-treated cells. The bioinformatics prediction revealed Bcl-2-like protein 11 (BCL2L11) and caspase-2 were two potential targets for miR-1247-3p, and functional assays confirmed that miR-1247-3p targeted both BCL2L11 and caspase-2 3' untranslated regions, which lead to the repressed expression of these genes. Silencing of BCL2L11 and caspase-2 both, respectively, counteracted the H9c2 cell injury caused by H/R treatment. Moreover, BCL2L11 and caspase-2 overexpression, respectively, impaired the protective effects of miR-1247-3p overexpression on H/R-treated H9c2 cells. The data in the present investigation revealed that miR-1247-3p restoration exhibited protective effects on H/R-induced cardiomyocyte injury through targeting BCL2L11 and caspase-2, implying that miR-1247-3p along with caspase-2/BCL2L11 signaling may provide novel sight for a better understating of I/R-induced myocardial damage. The role of miR-1247-3p might be further confirmed in animal models and clinical studies.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Z-Val-Asp(O-Me)-Val-Ala-Asp(O-Me) fluoromethyl ketone, powder, ≥95%