Merck
CLEAR: coverage-based limiting-cell experiment analysis for RNA-seq.
Journal of translational medicine (2020-02-11)
Logan A Walker, Michael G Sovic, Chi-Ling Chiang, Eileen Hu, Jiyeon K Denninger, Xi Chen, Elizabeth D Kirby, John C Byrd, Natarajan Muthusamy, Ralf Bundschuh, Pearlly Yan
ABSTRACT
Direct cDNA preamplification protocols developed for single-cell RNA-seq have enabled transcriptome profiling of precious clinical samples and rare cell populations without the need for sample pooling or RNA extraction. We term the use of single-cell chemistries for sequencing low numbers of cells limiting-cell RNA-seq (lcRNA-seq). Currently, there is no customized algorithm to select robust/low-noise transcripts from lcRNA-seq data for between-group comparisons. Herein, we present CLEAR, a workflow that identifies reliably quantifiable transcripts in lcRNA-seq data for differentially expressed genes (DEG) analysis. Total RNA obtained from primary chronic lymphocytic leukemia (CLL) CD5+ and CD5- cells were used to develop the CLEAR algorithm. Once established, the performance of CLEAR was evaluated with FACS-sorted cells enriched from mouse Dentate Gyrus (DG). When using CLEAR transcripts vs. using all transcripts in CLL samples, downstream analyses revealed a higher proportion of shared transcripts across three input amounts and improved principal component analysis (PCA) separation of the two cell types. In mouse DG samples, CLEAR identifies noisy transcripts and their removal improves PCA separation of the anticipated cell populations. In addition, CLEAR was applied to two publicly-available datasets to demonstrate its utility in lcRNA-seq data from other institutions. If imputation is applied to limit the effect of missing data points, CLEAR can also be used in large clinical trials and in single cell studies. lcRNA-seq coupled with CLEAR is widely used in our institution for profiling immune cells (circulating or tissue-infiltrating) for its transcript preservation characteristics. CLEAR fills an important niche in pre-processing lcRNA-seq data to facilitate transcriptome profiling and DEG analysis. We demonstrate the utility of CLEAR in analyzing rare cell populations in clinical samples and in murine neural DG region without sample pooling.
Product Description
Roche
Papain, from Carica papaya
Pricing and availability is not currently available.