Merck
  • Synthesis of novel carbazole based macrocyclic amides as potential antimicrobial agents.
Synthesis of novel carbazole based macrocyclic amides as potential antimicrobial agents.
European journal of medicinal chemistry (2008-09-10)
Perumal Rajakumar, Karuppannan Sekar, Vellasamy Shanmugaiah, Narayanasamy Mathivanan
ABSTRACT
A series of carbazole based macrocyclic diamides with thia and oxy linkages have been synthesized and the inhibitory activity of the cyclophane amides against human pathogenic bacteria and plant pathogenic fungi are documented. (S)-1,10-Bi-2-naphthol [(S)-BINOL] based chiral carbazolophane amide emerged as the most interesting compound in this series exhibiting excellent antibacterial and antifungal activities.
Sigma-Aldrich
Carbendazim, 97%
Supelco
Carbendazim, PESTANAL®, analytical standard
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)