Merck
  • Cellular growth inhibition by FK778 is linked to G1 arrest or S phase accumulation, dependent on the functional status of the retinoblastoma protein.
Cellular growth inhibition by FK778 is linked to G1 arrest or S phase accumulation, dependent on the functional status of the retinoblastoma protein.
International journal of molecular medicine (2009-02-13)
Karin Hoppe-Seyler, Kilian Weigand, Claudia Lohrey, Felix Hoppe-Seyler, Peter Sauer
ABSTRACT
The malononitrilamide FK778 is a novel immunosuppressive agent with antiproliferative activities. To gain insight into the molecular mechanism of FK778-mediated growth inhibition, we analyzed cells which differ in their p53 status and functionality of retinoblastoma protein (pRb). FK778 acted as a broad inhibitor of cell proliferation independent of the p53 or pRb status. However, the mechanism of FK778-mediated growth inhibition differed, leading either to cell cycle arrest in G1, or cell accumulation in S phase. This differential response was linked to the phosphorylation status of pRb. In addition, since FK778 was reported to exhibit antiviral activities, we analyzed the effect of FK778 on the growth stimulatory human papillomavirus (HPV)-16 and -18 E7 genes. Although growth of HPV-positive cells was strongly inhibited by FK778, we did not observe significant effects on viral E7 expression, indicating that the antiproliferative effect is not linked to an antiviral activity of FK778.
Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®