Merck
  • Role of long non-coding RNA in DEET- and fipronil-mediated alteration of transcripts associated with Phase I and Phase II xenobiotic metabolism in human primary hepatocytes.
Role of long non-coding RNA in DEET- and fipronil-mediated alteration of transcripts associated with Phase I and Phase II xenobiotic metabolism in human primary hepatocytes.
Pesticide biochemistry and physiology (2020-06-13)
Roger D Lawrie, Robert D Mitchell, Anirudh Dhammi, Andrew Wallace, Ernest Hodgson, R Michael Roe
ABSTRACT
Human exposure to environmental chemicals both individually and in combination occurs frequently world-wide most often with unknown consequences. Use of molecular approaches to aide in the assessment of risk involved in chemical exposure is a growing field in toxicology. In this study, we examined the impact of two environmental chemicals used in and around homes, the insect repellent DEET (N,N-diethyl-m-toluamide) and the phenylpyrazole insecticide fipronil (fluocyanobenpyrazole) on transcript levels of enzymes potentially involved in xenobiotic metabolism and on long non-coding RNAs (lncRNAs). Primary human hepatocytes were treated with these two chemicals both individually and in combination. Using RNA-Seq, we found that 10 major enzyme categories involved in phase 1 and phase 2 xenobiotic metabolism were significantly (α = 0.05) up- and down-regulated (i.e., 100 μM DEET-19 transcripts, 89% up and 11% down; 10 μM fipronil-52 transcripts, 53% up and 47% down; and 100 μM DEET +10 μM fipronil-69 transcripts, 43% up and 57% down). The altered genes were then mapped to the human genome and their proximity (within 1,000,000 bp) to lncRNAs examined. Unique proximities were discovered between altered lncRNA and altered P450s (CYP) and other enzymes (DEET, 2 CYP; Fipronil, 6 CYP and 15 other; and DEET + fipronil, 7 CYP and 21 other). Many of the altered P450 transcripts were in multiple clusters in the genome with proximal altered lncRNAs, suggesting a regulator function for the lncRNA. At the gene level there was high percent identity for lncRNAs near P450 clusters, but this relationship was not found at the transcript level. The role of these altered lncRNAs associated with xenobiotic induction, human diseases and chemical mixtures is discussed.
Product Description
SAFC
Williams′ Medium E, With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Gentamicin-Glutamine solution, with 200 mM L-glutamine and 5 mg gentamicin per mL in tissue culture grade water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture